4'-Fluoro[1,1'-biphenyl]-2-amine 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC50399-1G
PC50399-250MG
PC50399-5G
APOSPC50399-1GEA
252
EUR
APOSPC50399-1G
APOSPC50399-250MG
APOSPC50399-5G
4'-Fluoro[1,1'-biphenyl]-2-amine 95%
4'-Fluoro[1,1'-biphenyl]-2-amine
Formel:
C₁₂H₁₀FN Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424657 CAS-Nummer: 321-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...