2-Chlor-3-fluor-4-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-Fluoropyridine-4-Boronic Acid

PC5054-1G PC5054-500MG PC5054-5G
APOSPC5054-1GEA 89.9 EUR
APOSPC5054-1G APOSPC5054-500MG APOSPC5054-5G
2-Chlor-3-fluor-4-pyridinboronsäure
2-Chloro-3-fluoro-4-pyridineboronic acid
Formel: C₅H₄BClFNO₂
Molecular Weight: 175,35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04972403
CAS-Nummer: 937595-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR