2-Fluoro-5-(hydroxymethyl)benzeneboronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-5-(hydroxymethyl)phenyl)boronic acid

PC5058-1G PC5058-25G PC5058-5G
APOSPC5058-1GEA 21.9 EUR
APOSPC5058-1G APOSPC5058-25G APOSPC5058-5G
2-Fluoro-5-(hydroxymethyl)benzeneboronic acid
2-Fluoro-5-(hydroxymethyl)benzeneboronic acid
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
Schmelzpunkt: 64…76 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06656276
CAS-Nummer: 1072952-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR