2-Amino-3',4'-difluoracetophenon Hydrochlorid 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Difluorophenacylamine hydrochloride

PC5059-1G PC5059-5G
APOSPC5059-1GEA 618 EUR
APOSPC5059-1G APOSPC5059-5G
2-Amino-3',4'-difluoracetophenon Hydrochlorid 99%
2-Amino-3',4'-difluoracetophenon Hydrochlorid
Formel: C₈H₈ClF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD08532461
CAS-Nummer: 1209894-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR