Methylchlordifluoracetat 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC5070-100G
PC5070-25G
PC5070-5G
APOSPC5070-100GEA
143
EUR
APOSPC5070-100G
APOSPC5070-25G
APOSPC5070-5G
Methylchlordifluoracetat 97%
Methylchlordifluoracetat
Useful trifluoromethylating agent. Trifluoromethylation ofaryl and vinyl halides: Tetrahedron Letts.,32, 7689 (1991). Nitroaryl chlorides can also be trifluoromethylated.
Formel:
C₃H₃ClF₂O₂ Siedepunkt: 79…81 °C Dichte: 1,37 g/cm³ (25 °C) Flammpunkt: 19 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000775 CAS-Nummer: 1514-87-0 EINECS: 216-154-9 UN: 2924 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...