α,α-Difluorophenylacetic acid ethyl ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2,2-difluoro-2-phenylacetate, Ethyl difluoro(phenyl)acetate

PC5071-100G PC5071-25G
APOSPC5071-100GEA 493 EUR
APOSPC5071-100G APOSPC5071-25G
α,α-Difluorophenylacetic acid ethyl ester 98%
alpha,alpha-Difluorophenylacetic acid ethyl ester
Formel: C₁₀H₁₀F₂O₂
Molecular Weight: 200.19 g/mol
Siedepunkt: 70…72 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD08275296
CAS-Nummer: 2248-46-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR