2-Carboxy-4-fluorphenylboronsäure 97%

Lieferant: Apollo Scientific

Synonyme: 2-Borono-5-fluorobenzoic acid

PC5079-1G PC5079-5G
APOSPC5079-1GEA 123 EUR
APOSPC5079-1G APOSPC5079-5G
2-Carboxy-4-fluorphenylboronsäure 97%
2-Carboxy-4-fluorphenylboronsäure
Formel: C₇H₆BFO₄
Molecular Weight: 183,9296 g/mol
Schmelzpunkt: 183…186 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235078
CAS-Nummer: 874290-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR