1-Brom-2-fluor-6-nitrobenzol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Brom-1-fluor-3-nitrobenzol, 2-Brom-3-fluornitrobenzol
PC5083-5G
PC5083-1G
PC5083-25G
APOSPC5083-5GEA
31.9
EUR
APOSPC5083-5G
APOSPC5083-1G
APOSPC5083-25G
1-Brom-2-fluor-6-nitrobenzol
1-Bromo-2-fluoro-6-nitrobenzene
Formel:
C₆H₃BrFNO₂ Molecular Weight: 220 g/mol Schmelzpunkt: 42…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782080 CAS-Nummer: 59255-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...