2-Amino-6-fluorphenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoro-2-aminophenol

PC5087-10G PC5087-1G PC5087-5G
APOSPC5087-10GEA 79.9 EUR
APOSPC5087-10G APOSPC5087-1G APOSPC5087-5G
2-Amino-6-fluorphenol 97%
2-Amino-6-fluorphenol
Formel: C₆H₆FNO
Molecular Weight: 127.12 g/mol
Schmelzpunkt: 140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08532465
CAS-Nummer: 53981-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR