Methyl-6-fluorsalicylat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-fluoro-6-hydroxybenzoate

PC5092-1G PC5092-10G PC5092-5G
APOSPC5092-1GEA 23.2 EUR
APOSPC5092-1G APOSPC5092-10G APOSPC5092-5G
Methyl-6-fluorsalicylat 98%
Methyl-6-fluorsalicylat
Formel: C₈H₇FO₃
Molecular Weight: 170,1378 g/mol
Siedepunkt: 58…60 °C/0,3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00153207
CAS-Nummer: 72373-81-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR