4-Fluor-3-methoxybenzylbromid 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Bromomethyl)-1-fluoro-2-methoxybenzene
PC51006-1G
APOSPC51006-1GEA
36.4
EUR
APOSPC51006-1G
4-Fluor-3-methoxybenzylbromid 95+%
4-Fluor-3-methoxybenzylbromid
Formel:
C₈H₈BrFO Molecular Weight: 219,0509 g/mol Schmelzpunkt: 39…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972878 CAS-Nummer: 141080-73-1 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...