2-[(3-Fluorophenoxy)methyl]benzoic acid

Lieferant: Apollo Scientific

PC510103-1G PC510103-5G
APOSPC510103-1GEA 256 EUR
APOSPC510103-1G APOSPC510103-5G
2-[(3-Fluorophenoxy)methyl]benzoic acid
2-[(3-Fluorophenoxy)methyl]benzoic acid
Formel: C₁₄H₁₁FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12072102
CAS-Nummer: 114312-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR