4-Bromo-3-fluorophenylacetonitrile
Lieferant: Apollo Scientific
|
Gefahr
|
PC51077-1G
PC51077-25G
PC51077-5G
APOSPC51077-1GEA
58.2
EUR
APOSPC51077-1G
APOSPC51077-25G
APOSPC51077-5G
4-Bromo-3-fluorophenylacetonitrile
4-Bromo-3-fluorophenylacetonitrile
Formel:
C₈H₅BrFN Molecular Weight: 214.0300 g/mol Schmelzpunkt: 46…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756199 CAS-Nummer: 499983-13-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...