(2-Fluoro-6-nitrophenyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Fluoro-6-nitrobenzyl alcohol
PC51079-1G
PC51079-5G
APOSPC51079-1GEA
325
EUR
APOSPC51079-1G
APOSPC51079-5G
(2-Fluoro-6-nitrophenyl)methanol
(2-Fluoro-6-nitrophenyl)methanol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,13 g/mol Schmelzpunkt: 52…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110169 CAS-Nummer: 1643-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...