(2-Chloro-3-fluoropyridin-4-yl)methanol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-fluoro-4-(hydroxymethyl)pyridine

PC51084-1G PC51084-5G
APOSPC51084-1GEA 72.5 EUR
APOSPC51084-1G APOSPC51084-5G
(2-Chloro-3-fluoropyridin-4-yl)methanol 98%
(2-Chloro-3-fluoropyridin-4-yl)methanol
Formel: C₆H₅ClFNO
Molecular Weight: 161,56 g/mol
Schmelzpunkt: 94…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11045047
CAS-Nummer: 946127-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR