2-Chloro-3-fluoroisonicotinaldehyde 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-fluoropyridine-4-carbaldehyde

PC51085-1G PC51085-250MG PC51085-5G
APOSPC51085-1GEA 53.8 EUR
APOSPC51085-1G APOSPC51085-250MG APOSPC51085-5G
2-Chloro-3-fluoroisonicotinaldehyde 96%
2-Chloro-3-fluoroisonicotinaldehyde
Formel: C₆H₃ClFNO
Molecular Weight: 159.5455 g/mol
Siedepunkt: 82 °C/9mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD04972388
CAS-Nummer: 329794-28-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR