2,4-Difluoro-5-iodobenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC51099-1G
PC51099-5G
PC51099-10G
APOSPC51099-1GEA
0
EUR
APOSPC51099-1G
APOSPC51099-5G
APOSPC51099-10G
2,4-Difluoro-5-iodobenzoic acid
2,4-Difluoro-5-iodobenzoic acid
Formel:
C₇H₃F₂IO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226506 CAS-Nummer: 161531-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...