Methyl-5-fluor-2-methylbenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-fluoro-2-methylbenzoate
PC5114D-100G
PC5114D-25G
PC5114D-5G
APOSPC5114D-100GEA
87
EUR
APOSPC5114D-100G
APOSPC5114D-25G
APOSPC5114D-5G
Methyl-5-fluor-2-methylbenzoat
Methyl-5-fluor-2-methylbenzoat
Formel:
C₉H₉FO₂ Molecular Weight: 168,1700 g/mol Siedepunkt: 90…92 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221457 CAS-Nummer: 175278-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...