3-Bromo-2-fluorophenylboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Bromo-2-fluorophenyl)boronic acid, 3-Bromo-2-fluorobenzeneboronic acid
PC51169-1G
PC51169-25G
PC51169-5G
APOSPC51169-1GEA
21.9
EUR
APOSPC51169-1G
APOSPC51169-25G
APOSPC51169-5G
3-Bromo-2-fluorophenylboronic acid
3-Bromo-2-fluorophenylboronic acid
Formel:
C₆H₅BBrFO₂ Schmelzpunkt: 225…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664228 CAS-Nummer: 352535-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...