1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,6-Difluoro-3-nitrobenzotrifluoride

PC51179-1G PC51179-5G
APOSPC51179-1GEA 24.8 EUR
APOSPC51179-1G APOSPC51179-5G
1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene 98%
1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene
Formel: C₇H₂F₅NO₂
Molecular Weight: 227,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12924861
CAS-Nummer: 123973-36-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR