Methyl nonafluorobutyl ketone 98%

Lieferant: Apollo Scientific

Synonyme: Methyl Perfluorobutyl Ketone, 1H,1H,1H-Nonafluoro-2-hexanone, 3,3,4,4,5,5,6,6,6-Nonafluoro-2-hexanone

PC5118-5G PC5118-25G
APOSPC5118-5GEA 161 EUR
APOSPC5118-5G APOSPC5118-25G
Methyl nonafluorobutyl ketone 98%
Methyl nonafluorobutyl ketone
Formel: C₆H₃F₉O
Molecular Weight: 262.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153209
CAS-Nummer: 678-18-2
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR