(5-Amino-2-fluorophenyl)methanol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-2-fluorobenzyl alcohol
PC5127-1G
PC5127-5G
PC5127-25G
APOSPC5127-1GEA
21.9
EUR
APOSPC5127-1G
APOSPC5127-5G
APOSPC5127-25G
(5-Amino-2-fluorophenyl)methanol 99%
(5-Amino-2-fluorophenyl)methanol
Formel:
C₇H₈FNO Molecular Weight: 141,15 g/mol Schmelzpunkt: 97…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094257 CAS-Nummer: 84832-00-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...