Methyl tridecafluoroheptanoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Perfluoroheptanoic Acid Methyl Ester, Methyl Perfluoroheptanoate, Tridecafluoroheptanoic Acid Methyl Ester

PC5137-25G PC5137-5G PC5137-100G
APOSPC5137-25GEA 111 EUR
APOSPC5137-25G APOSPC5137-5G APOSPC5137-100G
Methyl tridecafluoroheptanoate 98%
Methyl tridecafluoroheptanoate
Formel: C₈H₃F₁₃O₂
Molecular Weight: 378,09 g/mol
Siedepunkt: 138…139 °C
Dichte: 1,483 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00153212
CAS-Nummer: 14312-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR