Ethyl-1-(trifluoromethyl)cyclopropanecarboxylate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 1-(trifluoromethyl)cyclopropanecarboxylate
PC51399-1G
APOSPC51399-1GEA
81.2
EUR
APOSPC51399-1G
Ethyl-1-(trifluoromethyl)cyclopropanecarboxylate 95%
Ethyl-1-(trifluoromethyl)cyclopropanecarboxylate
Formel:
C₇H₉F₃O₂ Molecular Weight: 182,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22199506 CAS-Nummer: 139229-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...