3-Amino-5-fluorpyridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoropyridin-3-amine

PC5151-1G PC5151-5G PC5151-25G
APOSPC5151-1GEA 21.9 EUR
APOSPC5151-1G APOSPC5151-5G APOSPC5151-25G
3-Amino-5-fluorpyridin 97%
3-Amino-5-fluoropyridine
Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
Schmelzpunkt: 85…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234073
CAS-Nummer: 210169-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR