2-Amino-5-chlor-3-nitrobenzotrifluorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-2-nitro-6-(trifluoromethyl)aniline
PC5161-1G
PC5161-5G
PC5161-25G
APOSPC5161-1GEA
21.9
EUR
APOSPC5161-1G
APOSPC5161-5G
APOSPC5161-25G
2-Amino-5-chlor-3-nitrobenzotrifluorid 98%
2-Amino-5-chlor-3-nitrobenzotrifluorid
Formel:
C₇H₄ClF₃N₂O₂ Molecular Weight: 240,5671 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094262 CAS-Nummer: 62924-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...