α-Methyl-2-trifluoromethylbenzyl alcohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[2-(Trifluoromethyl)phenyl]ethanol
PC5172M-25G
APOSPC5172M-25GEA
122
EUR
APOSPC5172M-25G
α-Methyl-2-trifluoromethylbenzyl alcohol 97%
alpha-Methyl-2-trifluoromethylbenzyl alcohol
Formel:
C₉H₉F₃O Molecular Weight: 190,17 g/mol Siedepunkt: 114…115 °C/2mmHg Schmelzpunkt: 40…42 °C Flammpunkt: 89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004513 CAS-Nummer: 79756-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...