α-Methyl-3-(trifluoromethyl)benzyl alcohol 97%
Lieferant: Apollo Scientific
PC5173-25G
PC5173-5G
APOSPC5173-25GEA
0
EUR
APOSPC5173-25G
APOSPC5173-5G
α-Methyl-3-(trifluoromethyl)benzyl alcohol 97%
alpha-Methyl-3-(trifluoromethyl)benzyl alcohol
Formel:
C₉H₉F₃O Siedepunkt: 89…91 °C/4mmHg Dichte: 1,249 g/cm³ (25 °C) Flammpunkt: 96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021868 CAS-Nummer: 454-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...