2-Fluoro-4-iodo-6-nitroaniline

Lieferant: Apollo Scientific
Achtung

PC5191-1G PC5191-100MG PC5191-250MG PC5191-500MG PC5191-2.5G PC5191-5G
APOSPC5191-1GEA 81.2 EUR
APOSPC5191-1G APOSPC5191-100MG APOSPC5191-250MG APOSPC5191-500MG APOSPC5191-2.5G APOSPC5191-5G
2-Fluoro-4-iodo-6-nitroaniline
2-Fluoro-4-iodo-6-nitroaniline
Formel: C₆H₄FIN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03094276
CAS-Nummer: 517920-73-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR