1,2-Diamino-3-fluoro-5-iodobenzene 97%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-5-iodobenzene-1,2-diamine

PC5192-1G PC5192-500MG
APOSPC5192-1GEA 434 EUR
APOSPC5192-1G APOSPC5192-500MG
1,2-Diamino-3-fluoro-5-iodobenzene 97%
1,2-Diamino-3-fluoro-5-iodobenzene
Formel: C₆H₆FIN₂
Molecular Weight: 252,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094277
CAS-Nummer: 517920-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR