3,5-Dimethyl-4-fluorbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC52041-1G
PC52041-5G
PC52041-10G
APOSPC52041-1GEA
31.9
EUR
APOSPC52041-1G
APOSPC52041-5G
APOSPC52041-10G
3,5-Dimethyl-4-fluorbenzonitril 98%
3,5-Dimethyl-4-fluorbenzonitril
Formel:
C₉H₈FN Molecular Weight: 149,1700 g/mol Schmelzpunkt: 88…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832248 CAS-Nummer: 867367-02-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...