Methyl-3-fluor-4-iodbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-fluoro-4-iodobenzoate

PC52047-1G PC52047-5G PC52047-25G
APOSPC52047-1GEA 49.3 EUR
APOSPC52047-1G APOSPC52047-5G APOSPC52047-25G
Methyl-3-fluor-4-iodbenzoat
Methyl-3-fluor-4-iodbenzoat
Formel: C₈H₆FIO₂
Molecular Weight: 280,0349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800728
CAS-Nummer: 1027513-46-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR