1-(2-Naphthoyl)-3,3,3-trifluoraceton 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione
PC5204M-100G
PC5204M-25G
PC5204M-500G
APOSPC5204M-100GEA
133
EUR
APOSPC5204M-100G
APOSPC5204M-25G
APOSPC5204M-500G
1-(2-Naphthoyl)-3,3,3-trifluoraceton 98%
1-(2-Naphthoyl)-3,3,3-trifluoraceton
Formel:
C₁₄H₉F₃O₂ Molecular Weight: 266,22 g/mol Schmelzpunkt: 70…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054501 CAS-Nummer: 893-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...