7-(Trifluoromethyl)quinoline 97%

Lieferant: Apollo Scientific

PC520509-10G PC520509-1G PC520509-5G
APOSPC520509-10GEA 757 EUR
APOSPC520509-10G APOSPC520509-1G APOSPC520509-5G
7-(Trifluoromethyl)quinoline 97%
7-(Trifluoromethyl)quinoline
Formel: C₁₀H₆F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00833760
CAS-Nummer: 325-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR