2-Amino-3,5,6-trifluoropyridine 97%
Lieferant: Apollo Scientific
PC520521-2.5G
PC520521-5G
APOSPC520521-2.5GEA
141
EUR
APOSPC520521-2.5G
APOSPC520521-5G
2-Amino-3,5,6-trifluoropyridine 97%
2-Amino-3,5,6-trifluoropyridine
Formel:
C₅H₃F₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001164 CAS-Nummer: 3534-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...