2-Chloro-5-fluoropyridine-3-carbaldehyde 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-fluoronicotinaldehyde

PC520523-5G
APOSPC520523-5GEA 180 EUR
APOSPC520523-5G
2-Chloro-5-fluoropyridine-3-carbaldehyde 97%
2-Chloro-5-fluoropyridine-3-carbaldehyde
Formel: C₆H₃ClFNO
Molecular Weight: 159.55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095247
CAS-Nummer: 851484-95-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR