(2-Chloro-5-fluoropyridin-3-yl)methanol 95%
Lieferant: Apollo Scientific
PC520524-10G
PC520524-1G
PC520524-5G
APOSPC520524-10GEA
1400
EUR
APOSPC520524-10G
APOSPC520524-1G
APOSPC520524-5G
(2-Chloro-5-fluoropyridin-3-yl)methanol 95%
(2-Chloro-5-fluoropyridin-3-yl)methanol
Formel:
C₆H₅ClFNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095313 CAS-Nummer: 870063-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...