4-(Trifluoromethylsulphony)phenylacetic acid 98%

Lieferant: Apollo Scientific

PC520538-25G PC520538-5G
APOSPC520538-25GEA 1400 EUR
APOSPC520538-25G APOSPC520538-5G
4-(Trifluoromethylsulphony)phenylacetic acid 98%
4-(Trifluoromethylsulphony)phenylacetic acid
Formel: C₉H₇F₃O₄S
Molecular Weight: 268,21 g/mol
Schmelzpunkt: 125…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04973012
CAS-Nummer: 1099597-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR