4-[2-(Trifluormethyl)phenoxy]piperidin 95%
Lieferant: Apollo Scientific
PC520542-10G
PC520542-1G
PC520542-5G
APOSPC520542-10GEA
1030
EUR
APOSPC520542-10G
APOSPC520542-1G
APOSPC520542-5G
4-[2-(Trifluormethyl)phenoxy]piperidin 95%
4-[2-(Trifluormethyl)phenoxy]piperidin
Formel:
C₁₂H₁₄F₃NO Molecular Weight: 245,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06248881 CAS-Nummer: 824390-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...