3,5-Dichlor-4-fluorpyridin 97%

Lieferant: Apollo Scientific

PC520543-1G PC520543-5G
APOSPC520543-1GEA 210 EUR
APOSPC520543-1G APOSPC520543-5G
3,5-Dichlor-4-fluorpyridin 97%
3,5-Dichloro-4-fluoropyridine
Formel: C₅H₂Cl₂FN
Molecular Weight: 165,9800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368620
CAS-Nummer: 916791-62-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR