1-(Difluoromethyl)-2-fluorobenzene 97%

Lieferant: Apollo Scientific

Synonyme: 2-Difluoromethyl-1-fluorobenzene

PC520545-25G PC520545-5G
APOSPC520545-25GEA 698 EUR
APOSPC520545-25G APOSPC520545-5G
1-(Difluoromethyl)-2-fluorobenzene 97%
1-(Difluoromethyl)-2-fluorobenzene
Formel: C₇H₅F₃
Molecular Weight: 146,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06657985
CAS-Nummer: 63878-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR