3-(Difluoromethyl)-1-fluorobenzene 97%
Lieferant: Apollo Scientific
PC520546-25G
PC520546-5G
APOSPC520546-25GEA
686
EUR
APOSPC520546-25G
APOSPC520546-5G
3-(Difluoromethyl)-1-fluorobenzene 97%
3-(Difluoromethyl)-1-fluorobenzene
Formel:
C₇H₅F₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657983 CAS-Nummer: 26029-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...