2-(Trifluoromethyl)quinoxaline 97%
Lieferant: Apollo Scientific
PC520550-10G
PC520550-5G
APOSPC520550-10GEA
582
EUR
APOSPC520550-10G
APOSPC520550-5G
2-(Trifluoromethyl)quinoxaline 97%
2-(Trifluoromethyl)quinoxaline
Formel:
C₉H₅F₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06656411 CAS-Nummer: 148853-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...