4-Brom-2-fluor-5-methylbenzonitril 95%

Lieferant: Apollo Scientific

PC520560-25G PC520560-5G
APOSPC520560-25GEA 0 EUR
APOSPC520560-25G APOSPC520560-5G
4-Brom-2-fluor-5-methylbenzonitril 95%
4-Brom-2-fluor-5-methylbenzonitril
Formel: C₈H₅BrFN
Molecular Weight: 214,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07782073
CAS-Nummer: 916792-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR