4-Fluoro-3-methylbenzotrifluoride 97%
Lieferant: Apollo Scientific
PC520564-10G
PC520564-5G
APOSPC520564-10GEA
919
EUR
APOSPC520564-10G
APOSPC520564-5G
4-Fluoro-3-methylbenzotrifluoride 97%
4-Fluoro-3-methylbenzotrifluoride
Formel:
C₈H₆F₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07778352 CAS-Nummer: 74483-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...