1-(Trifluormethyl)cyclohexancarbonsäure 97%

Lieferant: Apollo Scientific

PC520566-1G PC520566-500MG PC520566-5G
APOSPC520566-1GEA 231 EUR
APOSPC520566-1G APOSPC520566-500MG APOSPC520566-5G
1-(Trifluormethyl)cyclohexancarbonsäure 97%
1-(Trifluoromethyl)cyclohexanecarboxylic acid
Formel: C₈H₁₁F₃O₂
Molecular Weight: 196,700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445820
CAS-Nummer: 180918-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR