[4-(Trifluoromethyl)phenylsulphonyl]acetonitrile 95%

Lieferant: Apollo Scientific

PC520572-1G PC520572-5G
APOSPC520572-1GEA 118 EUR
APOSPC520572-1G APOSPC520572-5G
[4-(Trifluoromethyl)phenylsulphonyl]acetonitrile 95%
[4-(Trifluoromethyl)phenylsulphonyl]acetonitrile
Formel: C₉H₆F₃NO₂S
Molecular Weight: 249.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08741474
CAS-Nummer: 186405-37-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR