5-Chloro-2-methylbenzotrifluoride 97%
Lieferant: Apollo Scientific
PC520576-10G
PC520576-1G
PC520576-5G
APOSPC520576-10GEA
1040
EUR
APOSPC520576-10G
APOSPC520576-1G
APOSPC520576-5G
5-Chloro-2-methylbenzotrifluoride 97%
5-Chloro-2-methylbenzotrifluoride
Formel:
C₈H₆ClF₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037961 CAS-Nummer: 1839-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...