5-Fluoro-2-nitrobenzeneboronic acid 97+%

Lieferant: Apollo Scientific

Synonyme: (5-Fluoro-2-nitrophenyl)boronic acid

PC520577-1G PC520577-5G
APOSPC520577-1GEA 154 EUR
APOSPC520577-1G APOSPC520577-5G
5-Fluoro-2-nitrobenzeneboronic acid 97+%
5-Fluoro-2-nitrobenzeneboronic acid
Formel: C₆H₅BFNO₄
Molecular Weight: 184.92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037802
CAS-Nummer: 1329171-65-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR