3-Chloro-5-fluorobenzotrifluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-3-fluoro-5-(trifluoromethyl)benzene

PC52058-10G PC52058-1G PC52058-5G
APOSPC52058-10GEA 800 EUR
APOSPC52058-10G APOSPC52058-1G APOSPC52058-5G
3-Chloro-5-fluorobenzotrifluoride
3-Chloro-5-fluorobenzotrifluoride
Formel: C₇H₃ClF₄
Molecular Weight: 198,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11226526
CAS-Nummer: 1005764-23-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR